What medium can be used to maintain/manipulate neurons at ambient CO2?
We offer Gibco Hibernate Medium for this purpose. When supplemented with Gibco B-27 Supplement and Gibco GlutaMAX-1 Supplement, this medium can allow for the manipulation of neurons at ambient CO2 for at least 48 hours while retaining their viability, and preserve viable brain tissue for up to a month when stored at 4 degrees C. Two Hibernate media are offered:
- Gibco Hibernate-A Medium (Cat. No. A1247501) is formulated for postnatal neurons.
- Gibco Hibernate-E Medium (Cat. No. A1247601) is formulated for embryonic neurons.
These two media have the same formulation and differ only in osmolality.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
- Gibco Hibernate-A Medium (Cat. No. A1247501) is formulated for postnatal neurons.
- Gibco Hibernate-E Medium (Cat. No. A1247601) is formulated for embryonic neurons.
These two media have the same formulation and differ only in osmolality.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.