Thermo Scientific Chemicals

Hydantoïne, 99 %, Thermo Scientific Chemicals

Référence: A12486.22
100 g, Each
Thermo Scientific Chemicals

Hydantoïne, 99 %, Thermo Scientific Chemicals

Référence: A12486.22
100 g, Each
Quantité
Référence: A12486.22
Prix (EUR)
Quantité
-

Identifiants chimiques

CAS
461-72-3
IUPAC Name
imidazolidine-2,4-dione
Molecular Formula
C3H4N2O2
InChI Key
WJRBRSLFGCUECM-UHFFFAOYSA-N
SMILES
O=C1CNC(=O)N1
Form
Powder
Identification (FTIR)
Conforms
Appearance (Color)
White to cream or pale yellow
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Assay (HPLC)
≥98.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documentation et téléchargements

Certificats

    Foire aux questions (FAQ)

    Citations et références

    Search citations by name, author, journal title or abstract text

    Sécurité et manipulation


    Not Hazardous

    Not Hazardous