Thermo Scientific Chemicals

Ammonium iodide, 98+%, Thermo Scientific Chemicals

Catalog number: A12498.22
100 g, Each
Thermo Scientific Chemicals

Ammonium iodide, 98+%, Thermo Scientific Chemicals

Catalog number: A12498.22
100 g, Each
Quantity
Catalog number: A12498.22
also known as A12498-22
Price (USD)
Price: 61.80
Online price: 52.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12027-06-4
IUPAC Name
ammonium iodide
Molecular Formula
H4IN
InChI Key
UKFWSNCTAHXBQN-UHFFFAOYSA-N
SMILES
[NH4+].[I-]
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream to yellow or greenish-yellow
Assay (Titration ex Iodide)
≥98.0 to ≤102.0%

Description

Ammonium iodide is a laboratory reagent and used as a photographic chemicals. It has wide range of applications in chemical industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iodide is a laboratory reagent and used as a photographic chemicals. It has wide range of applications in chemical industries.

Solubility
Soluble in water (1770 mg/ml at 25°C), methanol (400 mg/ml), glycerol (667 mg/ml), alcohol (270 mg/ml), and ethanol.

Notes
Store at 4°C. Hande under dry protective gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text