Cloruro de tetrazolio azul, Thermo Scientific Chemicals
Cloruro de tetrazolio azul, Thermo Scientific Chemicals
Cloruro de tetrazolio azul, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cloruro de tetrazolio azul, Thermo Scientific Chemicals

CAS: 1871-22-3 | C40H36Cl2N8O2 | 731.68 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
25 g
Número de catálogo A12502.14
también denominado A12502-14
Precio (EUR)
612,00
Each
Cantidad:
25 g
Pedido a granel o personalizado
Precio (EUR)
612,00
Each
Identificadores químicos
CAS1871-22-3
IUPAC Namedihydrogen 2-[4'-(3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazol-2-yl)-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl]-3,5-diphenyl-2,3-dihydro-1H-1,2,3,4-tetrazole dichloride
Molecular FormulaC40H36Cl2N8O2
InChI KeyRCEHREKDVGHYAM-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].COC1=CC(=CC=C1N1NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=C(N2NC(=NN2C2=CC=CC=C2)C2=CC=CC=C2)C(OC)=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Pale yellow to yellow
FormPowder
Identification (FTIR)Conforms
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Blue Tetrazolium chloride is useful in the prepartion of DNA cation complexes and immunoadsorbents for anti-DNA antibodies. It is used in assay of reducing sugars. In combination with 4-nitrocatechol, tetrazolium blue chloride forms a ternary complex with molybdenum(VI) for extraction and spectrophotometric determination of molybdenum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El cloruro de tetrazolio azul resulta de utilidad en la preparación de complejos de cationes de ADN e inmunoadsorbentes para anticuerpos anti-ADN. Se utiliza en el ensayo de azúcares reductores. En combinación con el 4-nitrocatecol, el cloruro de tetrazolio azul forma un complejo ternario con molibdeno(VI) para la extracción y determinación espectrofotométrica de molibdeno.

Solubilidad
Soluble en agua caliente (5 mg/ml), metanol (50 mg/ml), etanol y cloroformo. Insoluble en acetona.

Notas
Estable en las condiciones de almacenamiento recomendadas. Incompatible con agentes oxidantes.
RUO – Research Use Only

General References:

  1. C.K.Jue; P.N.Lipke. Determination of reducing sugars in the nanomole range with tetrazolium blue chloride. Journal of Biochemical and Biophysical Methods. 1985, 11, (2-3)109-115
  2. Vincent A.DeBari; Mark A.Needle. Cation complexed DNA. A novel class of immunoadsorbents for anti-DNA antibodies. Journal of Immunological Methods. 1981, 40, (1)89-94
  3. Reagent for identification of reducing sugars.
  4. For use in the oxidation of steroids, see: J. Chem. Soc., Chem. Commun., 2687 (1988).