2,4-Dichlorobenzaldehyde, 98%
2,4-Dichlorobenzaldehyde, 98%
2,4-Dichlorobenzaldehyde, 98%
2,4-Dichlorobenzaldehyde, 98%
Thermo Scientific Chemicals

2,4-Dichlorobenzaldehyde, 98%

CAS: 874-42-0 | C7H4Cl2O | 175.008 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A12525.36
also known as A12525-36
Price (EUR)
109,00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)
109,00
Each
Add to cart
Chemical Identifiers
CAS874-42-0
IUPAC Name2,4-dichlorobenzaldehyde
Molecular FormulaC7H4Cl2O
InChI KeyYSFBEAASFUWWHU-UHFFFAOYSA-N
SMILESClC1=CC=C(C=O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or flakes
Assay (GC)≥97.5%
Free acid (titration)≤1.0%
Identification (FTIR)Conforms
Melting Point69.5-73.5?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only