Pentaacetato de beta-D-glucosa, 98 %, Thermo Scientific Chemicals
Pentaacetato de beta-D-glucosa, 98 %, Thermo Scientific Chemicals
Pentaacetato de beta-D-glucosa, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pentaacetato de beta-D-glucosa, 98 %, Thermo Scientific Chemicals

CAS: 604-69-3 | C16H22O11 | 390.341 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
50 g
250 g
1000 g
Número de catálogo A12526.18
también denominado A12526-18
Precio (EUR)
40,00
Each
Cantidad:
50 g
Pedido a granel o personalizado
Precio (EUR)
40,00
Each
Identificadores químicos
CAS604-69-3
IUPAC Name[3,4,5,6-tetrakis(acetyloxy)oxan-2-yl]methyl acetate
Molecular FormulaC16H22O11
InChI KeyLPTITAGPBXDDGR-UHFFFAOYNA-N
SMILESCC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay (GC)≥97.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤0.5%
Ver más
β-D-Glucose Pentaacetate is used in biochemical reaction and also used as an active pharmaceutical intermediate. Further, it is used to stimulate insulin release in rat pancreatic islets.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El Pentaacetati de β-D-glucosa se utiliza en la reacción bioquímica y como un intermediario farmacéutico activo. Además, se utiliza para estimular la liberación de insulina en los islotes pancreáticos de rata.

Solubilidad
Soluble en cloroformo y metanol. Insoluble en agua.

Notas
Almacenar en un lugar fresco. Incompatible con agentes oxidantes fuertes.
RUO – Research Use Only

General References:

  1. Ali, T. H.; Tajuddin, H. A. B.; Hussen, R. S. D.; Heidelberg, T. Increased Emulsion Stability for Reverse Y-Shaped Sugar-Based Surfactants. J. Surfactants Deterg. 2015, 18 (5), 881-886.
  2. Lülsdorf, N.; Pitzler, C.; Biggel, M.; Martinez, R.; Vojcic, L.; Schwaneberg, U. A flow cytometer-based whole cell screening toolbox for directed hydrolase evolution through fluorescent hydrogels. Chem. Commun. 2015, 51 (41), 8679-8682.