4-Chlorophenylacetyl chloride, 98%
4-Chlorophenylacetyl chloride, 98%
4-Chlorophenylacetyl chloride, 98%
Thermo Scientific Chemicals

4-Chlorophenylacetyl chloride, 98%

CAS: 25026-34-0 | C8H6Cl2O | 189.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A12530.09
also known as A12530-09
Price (EUR)
43,90
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
43,90
Each
Chemical Identifiers
CAS25026-34-0
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.5490-1.5565 @ 20?C
FormLiquid
Appearance (Color)Clear yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only