Thermo Scientific Chemicals

4-Chlorobenzenesulfinic acid sodium salt hydrate, 97%, Thermo Scientific Chemicals

Catalog number: A12583.14
25 g, Each
Thermo Scientific Chemicals

4-Chlorobenzenesulfinic acid sodium salt hydrate, 97%, Thermo Scientific Chemicals

Catalog number: A12583.14
25 g, Each
Quantity
Catalog number: A12583.14
also known as A12583-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
14752-66-0
IUPAC Name
sodium 4-chlorobenzene-1-sulfinate
Molecular Formula
C6H4ClNaO2S
InChI Key
JFXAUUFCZJYLJF-UHFFFAOYSA-M
SMILES
[Na+].[O-]S(=O)C1=CC=C(Cl)C=C1
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
≤18.48%
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥96.0 to ≤104.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text