2,4-Dichloro-6-methylaniline, 97%
2,4-Dichloro-6-methylaniline, 97%
2,4-Dichloro-6-methylaniline, 97%
2,4-Dichloro-6-methylaniline, 97%
Thermo Scientific Chemicals

2,4-Dichloro-6-methylaniline, 97%

CAS: 30273-00-8 | C7H7Cl2N | 176.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12591.14
also known as A12591-14
Price (EUR)
197,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
197,00
Each
Add to cart
Chemical Identifiers
CAS30273-00-8
IUPAC Name2,4-dichloro-6-methylaniline
Molecular FormulaC7H7Cl2N
InChI KeyUAISVUGQLKXPFF-UHFFFAOYSA-N
SMILESCC1=CC(Cl)=CC(Cl)=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)White to pale cream
FormPowder
Melting Point (clear melt)38.0-47.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only