Thermo Scientific Chemicals

4-Aminobenzamide, 98+%, Thermo Scientific Chemicals

Catalog number: A12594.14
25 g, Each
Thermo Scientific Chemicals

4-Aminobenzamide, 98+%, Thermo Scientific Chemicals

Catalog number: A12594.14
25 g, Each
Quantity
Catalog number: A12594.14
also known as A12594-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
2835-68-9
IUPAC Name
4-aminobenzamide
Molecular Formula
C7H8N2O
InChI Key
QIKYZXDTTPVVAC-UHFFFAOYSA-N
SMILES
NC(=O)C1=CC=C(N)C=C1
Melting Point (clear melt)
179-185?C
Appearance (Color)
White to cream
Form
Powder or crystalline powder
Assay (HPLC)
≥98.0%

Description

4-Aminobenzamide was used as a poly(ADP-ribose)polymerase (PADPRP) inhhibitor to study the death of target cells by cytotoxic effector cells using the nuclear enzyme poly(ADP-ribose)polymerase (PADPRP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzamide was used as a poly(ADP-ribose)polymerase (PADPRP) inhhibitor to study the death of target cells by cytotoxic effector cells using the nuclear enzyme poly(ADP-ribose)polymerase (PADPRP).

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text