4-Chloro-3-nitrobenzonitrile, 98%
4-Chloro-3-nitrobenzonitrile, 98%
4-Chloro-3-nitrobenzonitrile, 98%
4-Chloro-3-nitrobenzonitrile, 98%
Thermo Scientific Chemicals

4-Chloro-3-nitrobenzonitrile, 98%

CAS: 939-80-0 | C7H3ClN2O2 | 182.563 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12648.06
also known as A12648-06
Price (EUR)
43,70
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
43,70
Each
Add to cart
Chemical Identifiers
CAS939-80-0
IUPAC Name4-chloro-3-nitrobenzonitrile
Molecular FormulaC7H3ClN2O2
InChI KeyXBLPHYSLHRGMNW-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(=CC=C1Cl)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)97.5-103.5?C
Appearance (Color)White to pale cream to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only