2-n-Hexylthiophene, 98%
2-n-Hexylthiophene, 98%
2-n-Hexylthiophene, 98%
Thermo Scientific Chemicals

2-n-Hexylthiophene, 98%

CAS: 18794-77-9 | C10H16S | 168.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A12652.09
also known as A12652-09
Price (TWD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS18794-77-9
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.4940-1.4980 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥97.5%
2-n-Hexylthiophene is used as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-n-Hexylthiophene is used as a important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Soluble in most organic solvents and ethanol Soluble . Insoluble in water.

Notes
Store in cool dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. William Barford; David G. Lidzey; Dmitry V. Makhov; and Anthony J. H. Meijer. Exciton localization in disordered poly(3-hexylthiophene). J. Chem. Phys. 2010, 133044504.
  2. M. A. V. Ribeiro da Silva; Ana Filipa L. O. M. Santos. Thermochemical study of 2- and 3-alkyl substituted thiophenes. Journal of Thermal Analysis and Calorimetry. 2007, 88(1), 7-17.