3,4-(Methylenedioxy)benzyl alcohol, 98%
3,4-(Methylenedioxy)benzyl alcohol, 98%
3,4-(Methylenedioxy)benzyl alcohol, 98%
Thermo Scientific Chemicals

3,4-(Methylenedioxy)benzyl alcohol, 98%

CAS: 495-76-1 | C8H8O3 | 152.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A12661.14
also known as A12661-14
Price (EUR)
29,40
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
29,40
Each
Chemical Identifiers
CAS495-76-1
IUPAC Name(2H-1,3-benzodioxol-5-yl)methanol
Molecular FormulaC8H8O3
InChI KeyBHUIUXNAPJIDOG-UHFFFAOYSA-N
SMILESOCC1=CC=C2OCOC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
FormCrystals or powder or crystalline powder
Melting Point (clear melt)49.0-55.0?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only