2-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol
2-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol
2-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol
Thermo Scientific Chemicals

2-Methylstyrene, 98%, stab. with 0.1% 4-tert-butylcatechol

CAS: 611-15-4 | C9H10 | 118.179 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12676.06
also known as A12676-06
Price (USD)
145.65
Online Exclusive
162.00
Save 16.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
145.65
Online Exclusive
162.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS611-15-4
IUPAC Name1-ethenyl-2-methylbenzene
Molecular FormulaC9H10
InChI KeyNVZWEEGUWXZOKI-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥95.0%
Refractive Index1.5410-1.5460 @ 20°C
SolutionClear: 10% in Diethyl ether
View more
2-Methylstyrene is a monomer used in the preparation of poly(2-methylstyrene). It is also used as an intermediate in synthetic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylstyrene is a monomer used in the preparation of poly(2-methylstyrene). It is also used as an intermediate in synthetic chemistry.

Solubility
Slightly miscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Wang, M.; Gao, B.; Huang, H. Catalytic nucleophilic addition of olefinic C-H bond to alpha, beta-unsaturated-gama-lactams. Tetrahedron Lett. 2015, 56 (41), 5533-5536.
  2. Chen, J.; Hua, L.; Chen, C.; Guo, L.; Zhang, R.; Chen, A.; Xiu, Y.; Liu, X.; Hou, Z. Temperature-Dependent Immobilization of a Tungsten Peroxo Complex That Catalyzes the Hydroxymethoxylation of Olefins. ChemPlusChem 2015, 80 (6), 1029-1037.