Thermo Scientific Chemicals

Nicotinic acid, 99%, Thermo Scientific Chemicals

Catalog number: A12683.0I
5000 g, Each
Thermo Scientific Chemicals

Nicotinic acid, 99%, Thermo Scientific Chemicals

Catalog number: A12683.0I
5000 g, Each
Quantity
Catalog number: A12683.0I
also known as A12683-0I
Price (USD)
Price: 336.00
Online price: 285.65
Your price:
Quantity
-

Chemical Identifiers

CAS
59-67-6
IUPAC Name
pyridine-3-carboxylic acid
Molecular Formula
C6H5NO2
InChI Key
PVNIIMVLHYAWGP-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CN=C1
Assay (Silylated GC)
≥98.5% (UK-sourced material)
Comment
Material sourced in UK and US
Melting Point (clear melt)
234.0-241.0°C
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5% (UK-sourced material)

Description

Vitamin of the B complex with hypolipidemic properties occurring in various animal and plant tissues. Required by the body for the formation of coenzymes NAD and NADP.Nicotinic acid is used as an antidyslipidemic agent. It is also used to improve all lipoproteins well above the vitamin requirement and lowers the total cholesterol, bad LDL-cholesterol, and triglyceride levels. Further, it is used as a food additive in food industries, cosmetics and pharmaceuticals. In addition to this, it is used in medication, nutrient, enriched flours and in electroplating baths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. Slightly soluble in ethanol and ethyl ether. Insoluble in most lipid solvents.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text