3-Amino-5-phenyl-1H-pyrazole, 98%
3-Amino-5-phenyl-1H-pyrazole, 98%
3-Amino-5-phenyl-1H-pyrazole, 98%
Thermo Scientific Chemicals

3-Amino-5-phenyl-1H-pyrazole, 98%

CAS: 1572-10-7 | C9H9N3 | 159.192 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12701.14
also known as A12701-14
Price (EUR)
431,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
431,00
Each
Chemical Identifiers
CAS1572-10-7
IUPAC Name5-phenyl-1H-pyrazol-3-amine
Molecular FormulaC9H9N3
InChI KeyPWSZRRFDVPMZGM-UHFFFAOYSA-N
SMILESNC1=NNC(=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)123.0-130.0°C
Appearance (Color)White to dark cream to pale brown
Assay (GC)≥97.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only