3',5'-Di-tert-butyl-4'-hydroxyacetophenone, 98%
3',5'-Di-tert-butyl-4'-hydroxyacetophenone, 98%
3',5'-Di-tert-butyl-4'-hydroxyacetophenone, 98%
3',5'-Di-tert-butyl-4'-hydroxyacetophenone, 98%
Thermo Scientific Chemicals

3',5'-Di-tert-butyl-4'-hydroxyacetophenone, 98%

CAS: 14035-33-7 | C16H24O2 | 248.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12722.06
also known as A12722-06
Price (TWD)
-
Request A Quote
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS14035-33-7
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)145.5-151.5?C
Appearance (Color)White to cream to pale brown or pink
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only