1,3-Dibromobenzene, 97+%
1,3-Dibromobenzene, 97+%
1,3-Dibromobenzene, 97+%
Thermo Scientific Chemicals

1,3-Dibromobenzene, 97+%

CAS: 108-36-1 | C6H4Br2 | 235.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A12723.22
also known as A12723-22
Price (EUR)
160,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
160,00
Each
Chemical Identifiers
CAS108-36-1
IUPAC Name1,3-dibromobenzene
Molecular FormulaC6H4Br2
InChI KeyJSRLURSZEMLAFO-UHFFFAOYSA-N
SMILESBrC1=CC(Br)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.0%
Refractive Index1.6055-1.6105 @ 20?C
1,3-Dibromobenzene is used in Suzuki coupling reaction for the preparation of [n]metacyclophanes. It is used to prepare 5-(3-bromo-phenyl)-2,4-di-tert-butoxy-pyrimidine using Pd(0) (PPh3)4 as a reagent. Further, it undergoes solid-supported potassium fluoride-aluminum oxide palladium-catalyzed polyarylation reaction with phenyl boronic acid to get conjugated polyaryls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dibromobenzene is used in Suzuki coupling reaction for the preparation of [n]metacyclophanes. It is used to prepare 5-(3-bromo-phenyl)-2,4-di-tert-butoxy-pyrimidine using Pd(0) (PPh3)4 as a reagent. Further, it undergoes solid-supported potassium fluoride-aluminum oxide palladium-catalyzed polyarylation reaction with phenyl boronic acid to get conjugated polyaryls.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. In the presence of NiCl2, cross-coupling with aryl Grignards gives m-terphenyls: Synthesis, 147 (1990).
  2. Monte, M. J. S.; Almeida, A. R. R. P.; Liebman, J. F. Prediction of enthalpy and standard Gibbs energy of vaporization of haloaromatics from atomic properties. Chemosphere 2015, 138, 478-485.
  3. Zhang, S.; Xu, Q. L.; Xia, J. C.; Jing, Y. M.; Zheng, Y. X.; Zuo, J. L. Highly efficient yellow phosphorescent organic light-emitting diodes with novel phosphine oxide-based bipolar host materials. J. Mater. Chem. C 2015, 3 (43), 11540-11547.