Cyclohexylacetone, 95%
Cyclohexylacetone, 95%
Cyclohexylacetone, 95%
Thermo Scientific Chemicals

Cyclohexylacetone, 95%

CAS: 103-78-6 | C9H16O | 140.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A12748.14
also known as A12748-14
Price (EUR)
582,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
582,00
Each
Chemical Identifiers
CAS103-78-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange or red-brown
Assay (GC)≥94.0%
Refractive Index1.4500-1.4560 @ 20°C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only