Thermo Scientific Chemicals

Tetraphenylcyclopentadienone, 98%, Thermo Scientific Chemicals

Catalog number: A12756.22
100 g, Each
Thermo Scientific Chemicals

Tetraphenylcyclopentadienone, 98%, Thermo Scientific Chemicals

Catalog number: A12756.22
100 g, Each
Quantity
Catalog number: A12756.22
also known as A12756-22
Price (USD)
Price: 300.00
Online price: 259.65
Your price:
Quantity
-

Chemical Identifiers

CAS
479-33-4
IUPAC Name
tetraphenylcyclopenta-2,4-dien-1-one
Molecular Formula
C29H20O
InChI Key
PLGPSDNOLCVGSS-UHFFFAOYSA-N
SMILES
O=C1C(=C(C(=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Black
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥97.5%
Melting Point (clear melt)
216.0-222.0?C

Description

It is an easily made building block for many organic and organometallic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an easily made building block for many organic and organometallic compounds.

Solubility
Soluble in Benzene. Insoluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text