2,6-Dimethyl-4-pyrone, 99%
2,6-Dimethyl-4-pyrone, 99%
2,6-Dimethyl-4-pyrone, 99%
2,6-Dimethyl-4-pyrone, 99%
Thermo Scientific Chemicals

2,6-Dimethyl-4-pyrone, 99%

CAS: 1004-36-0 | C7H8O2 | 124.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A12799.30
also known as A12799-30
Price (EUR)
525,00
Each
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (EUR)
525,00
Each
Add to cart
Chemical Identifiers
CAS1004-36-0
IUPAC Name2,6-dimethyl-4H-pyran-4-one
Molecular FormulaC7H8O2
InChI KeyVSYFZULSKMFUJJ-UHFFFAOYSA-N
SMILESCC1=CC(=O)C=C(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)131.0-137.0?C
Appearance (Color)White to cream to pale brown
Assay (GC)≥98.5%
2,6-Dimethyl-4-pyrone is used as a chemical and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethyl-4-pyrone is used as a chemical and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressure.
RUO – Research Use Only

General References:

  1. LL Woods. Some further reactions of 2, 6-dimethyl-4-pyrone. Journal of the American Chemical Society. 195880, (6) , 1440-1442.
  2. D Cook. Infrared spectra of 2, 6-dimethyl-4-pyrone complexes. Journal of Chemistry. 196139, (6) , 1184-1189.