Bromoethane, 98%
Bromoethane, 98%
Bromoethane, 98%
Thermo Scientific Chemicals

Bromoethane, 98%

CAS: 74-96-4 | C2H5Br | 108.966 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A12804.0E
also known as A12804-0E
Price (USD)
126.65
Special offer
Online exclusive
Ends: 30-Jun-2026
149.00
Save 22.35 (15%)
Each
Quantity:
2500 g
Request bulk or custom format
Price (USD)
126.65
Special offer
Online exclusive
Ends: 30-Jun-2026
149.00
Save 22.35 (15%)
Each
Chemical Identifiers
CAS74-96-4
IUPAC Namebromoethane
Molecular FormulaC2H5Br
InChI KeyRDHPKYGYEGBMSE-UHFFFAOYSA-N
SMILESCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.4225-1.4265 @ 20?C
Bromoethane is used in organic synthesis for the introduction of ethyl group. It is widely used as solvent, as an anesthetic in medicine and as a refrigerant. It is an important component of antiknock fluid in gasoline. It is also also used in the manufacture of pharmaceuticals like vitamin A as well as in the manufacture of flame retardants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromoethane is used in organic synthesis for the introduction of ethyl group. It is widely used as solvent, as an anesthetic in medicine and as a refrigerant. It is an important component of antiknock fluid in gasoline. It is also also used in the manufacture of pharmaceuticals like vitamin A as well as in the manufacture of flame retardants.

Solubility
Miscible with ethanol, ether, chloroform and organic solvents.

Notes
Light sensitive. Store in cool place. Incompatible with strong bases, strong oxidizing agents, magnesium, potassium, calcium, sodium oxides, aluminum, zinc and alkali metals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Sarzyński, D.; Gola, A. A.; Dryś, A.; Brudnik, K.; Jodkowski, J. T. Kinetic study of the reaction of chlorine atoms with bromoethane and D-bromoethane in the gas phase. Chem. Phys. Lett. 2011, 509 (4-6), 114-118.
  2. Zhu, J.; Zhang, Y.; Tian, M.; Liu, J. Fabrication of a Mixed Matrix Membrane with in Situ Synthesized Quaternized Polyethylenimine Nanoparticles for Dye Purification and Reuse. ACS Sustainable Chem. Eng. 2015, 3 (4), 690-701.