Tetramethylammonium iodide, 99%
Tetramethylammonium iodide, 99%
Tetramethylammonium iodide, 99%
Thermo Scientific Chemicals

Tetramethylammonium iodide, 99%

CAS: 75-58-1 | C4H12IN | 201.05 g/mol
製品番号(カタログ番号)数量
A12811.22
または、製品番号A12811-22
100 g
製品番号(カタログ番号) A12811.22
または、製品番号A12811-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS75-58-1
IUPAC Nametetramethylazanium iodide
Molecular FormulaC4H12IN
InChI KeyRXMRGBVLCSYIBO-UHFFFAOYSA-M
SMILES[I-].C[N+](C)(C)C
さらに表示
Assay (Titration ex Iodide)≥98.5% to ≤101.5% (non-U.S. specification)
Assay≥98.5% (NMR) (U.S. specification)
Appearance (Color)White to pale yellow
FormPowder or crystalline powder
CommentSpecification differs from U.S. and non-U.S. material where indicated
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only