3-Methoxyphenol, 97%
3-Methoxyphenol, 97%
3-Methoxyphenol, 97%
Thermo Scientific Chemicals

3-Methoxyphenol, 97%

CAS: 150-19-6 | C7H8O2 | 124.139 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
500 g
產品號碼 A12826.22
亦稱為 A12826-22
價格 (TWD)
-
Quantity:
100 g
Request bulk or custom format
化學識別
CAS150-19-6
IUPAC Name3-methoxyphenol
Molecular FormulaC7H8O2
InChI KeyASHGTJPOSUFTGB-UHFFFAOYSA-N
SMILESCOC1=CC=CC(O)=C1
檢視更多
規格Specification Sheet規格表
Refractive Index1.5490-1.5540 @ 20?C
Appearance (Color)Clear colorless to yellow to orange to red to brown or pink
Assay (GC)≥96.0%
FormLiquid
It is used in the synthesis of C(4) symmetric calix[4]resorcinarene, 2-nitroso-5-methoxyphenol and 6-methoxy-2(3H)-benzoxazolone. It is also used as a flavoring agent. They are also used are used to make antioxidants and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of C(4) symmetric calix[4]resorcinarene, 2-nitroso-5-methoxyphenol and 6-methoxy-2(3H)-benzoxazolone. It is also used as a flavoring agent. They are also used are used to make antioxidants and pharmaceuticals.

Solubility
Partly soluble in water.

Notes
Store away from oxidizing agents, acids, bases, acid chlorides and acid anhydrides. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources
RUO – Research Use Only

General References:

  1. McIldowie MJ.; Mocerino M.; Skelton BW, et al.Facile lewis acid catalyzed synthesis of C(4) symmetric resorcinarenes.Org. Lett.,2000,2(24), 3869-71.
  2. Maleski RJ. Improved procedures for the preparation of 2-nitro-5-methoxyphenol and 6-methoxy-2 (3H)-benzoxazolone from 3-methoxyphenol.Synth. Commun.,1993,23(3), 343-8.