CTS™ DPBS, without calcium chloride, without magnesium chloride, 1 L - FAQs
CTS™ DPBS, without calcium chloride, without magnesium chloride, 1 L - FAQs
View additional product information for CTS™ DPBS, without calcium chloride, without magnesium chloride - FAQs (A1285601, A1285602)
3 product FAQs found
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
Can I get a specific value for the conductivity for your DPBS products?
Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Are your DPBS products DNase/RNase-free? Can I use them for RNA work?
Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.