CTS™ GlutaMAX™-I Supplement, 100 mL bottle - FAQs
CTS™ GlutaMAX™-I Supplement, 100 mL bottle - FAQs
View additional product information for CTS™ GlutaMAX™ Supplement - FAQs (A4737003, A4737001, A1286001)
4 product FAQs found
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
What is the difference between GlutaMAX Supplement (Cat. No. 35050061), CTS GlutaMAX-I Supplement (Cat. No. A1286001), and CTS GlutaMAX Supplement (Cat. No. A4737001)?
The formulation and manufacturing process are the same for all three products. The main difference is in the “intended use” statement. CTS GlutaMAX-I Supplement (Cat. No. A1286001) and CTS GlutaMAX Supplement (Cat. No. A4737001) have an intended use statement of “For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals”. GlutaMAX Supplement (Cat. No. 35050061) has an intended use statement of “For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals”. The catalog number to use will depend on the specific application.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Will depletion, absence, or breakdown of essential growth-promoting components such as glutamine or growth factors reduce the growth rate of my culture?
Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax I or II for glutamine in the medium to prevent glutamine exhaustion.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Will the media formulations containing GlutaMAX supplement change with respect to L-glutamine content?
In all media containing GlutaMAX supplement dipeptides as a substitute for L-glutamine, concentration is equimolar with the L-glutamine in the original formulation.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.