Invitrogen™

BacMam ERK2 [pThr185/pTyr187] Cellular Assay Kit

Catalog number:  A12888

 Related applications:

Pharma & Biopharma

| Target & Lead Identification & Validation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The BacMam ERK2 [pThr185⁄pTyr187] Cellular Assay Kit is a high-throughput screening (HTS) compatible cellular immunoassay for the measuring of phosphorylation of GFP-ERK2. The BacMam ERK2 Cellular Assay Kit can be used with numerous transformed cell lines, primary cells, and stem cells, and allows for the interrogation of ERK2, either as a kinase or GPCR readout.

• Choose cell types that are relevant to your needs
• Get results in weeks instead of months
• Perform Assays on Your Schedule

Use Cell Types Relevant to Your Needs
Understanding signaling in physiologically relevant cell backgrounds is extremely important when predicting the real effects of compounds in humans prior to clinical trials. With the BacMam ERK2 [pThr185⁄pTyr187] Cellular Assay Kit you can choose the appropriate cell type for your research needs, enabling you to design your own experiments, and to reach your research goals faster.

BacMam ERK2 Reagent Compatible Cells
The BacMam reagent included in the kit has been used to successfully transduce numerous transformed cell lines (e.g. U2-OS, HEK 293, and HeLa), primary cells (e.g., fibroblasts, hepatocytes, cardiovascular cells and epithelial cells), and stem cells (e.g. neuronal and mesenchymal) at high transduction efficiency and low levels of cytotoxicity. However, cells of hematopoietic origin show consistently poor transduction by BacMam technology, and are not recommended for use with the current technology.

Develop Assays Faster
The BacMam platform enables transient target expression allowing for reduced time to an HTS ready assay versus traditional stable cell line creation. Traditional stable cell line development requires months of culturing, optimization, and cloning of your cell line. This requires considerable time and resources. With BacMam ERK2 a user can get results in weeks versus months.

Cells Ready When You Are
BacMam Erk2 Reagent is ideal to transduce large quantities of cells in batch mode; transduced cells can be stored frozen in aliquots for later use. This saves you the delays associated with culturing cells. The cells are ready whenever you are. Cells can be assayed typically within hours of thawing, providing assay-ready cells when you need them, with no loss of activity.

For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Gene Symbol: ERK2
Cell State: Cells Not Included
Delivery Method: Baculovirus (BacMam System)
Detection Method: Fluorescent
Druggable Target: GPCRs, Kinases
Product Size: 1 kit
Readout: End Point
System: LanthaScreen®
Technique: TR-FRET

Contents & storage

Kit contains Terbium-labeled anti-ERK2 [pThr185/pTyr187] antibody, BacMam ERK2 Virus, 6X Lysis Buffer, and BacMam Enhancer Solution. Store as indicated in handling documentation.

Documents

Manuals & protocols