BacMam Akt [pThr308] Cellular Assay Kit
Product Image
Thermo Scientific™

BacMam Akt [pThr308] Cellular Assay Kit

The combination of baculovirus-mediated gene delivery (BacMam) with LanthaScreen™ Cellular Assay technology enables a platform for the analysis of specificRead more
Have Questions?
Catalog number A12900
Price (USD)/ Each
3,315.00
-
Add to cart
Quantity:
1 kit
Price (USD)/ Each
3,315.00
Add to cart
BacMam Akt [pThr308] Cellular Assay Kit
Catalog numberA12900
Price (USD)/ Each
3,315.00
-
Add to cart

The combination of baculovirus-mediated gene delivery (BacMam) with LanthaScreen™ Cellular Assay technology enables a platform for the analysis of specific posttranslational modifications of various substrates including AKT. BacMam provides a convenient genetic delivery tool for a GFP-AKT fusion protein in the cell line of interest. This kit describes an HTS-compatible cellular immunoassay measuring phosphorylation of GFP-AKT at Thr308.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Assay EntryCell-based terbium modification activity (transient), Cell-based terbium phospho-specific activity (transient)
Cell StateCells Not Included
Detection MethodFluorescence
For Use With (Application)TR-FRET
Human Protein ClassKinases, Cell Signalling
Product LineLanthaScreen™
Quantity1 kit
ReadoutEnd Point
Reporter GeneGFP (EmGFP)
Target EntryAKT1
System TypeLanthaScreen™
Druggable TargetKinases
Unit SizeEach
Contents & Storage
Kit contains Terbium-labeled anti-AKT antibody, BacMam AKT Virus, and 6X Lysis Buffer. Store as indicated in handling documentation.