4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzenesulfonyl chloride, 98%

CAS: 2991-42-6 | C7H4ClF3O2S | 244.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A12907.06
also known as A12907-06
Price (EUR)
82,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
82,00
Each
Add to cart
Chemical Identifiers
CAS2991-42-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow or pale cream
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥97.5%
Melting Point (clear melt)28-34?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only