Thermo Scientific Chemicals

Syringaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: A12933.22
100 g, Each
Thermo Scientific Chemicals

Syringaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: A12933.22
100 g, Each
Quantity
Catalog number: A12933.22
also known as A12933-22
Price (USD)
Price: 217.00
Online price: 189.65
Your price:
Quantity
-

Chemical Identifiers

CAS
134-96-3
IUPAC Name
4-hydroxy-3,5-dimethoxybenzaldehyde
Molecular Formula
C9H10O4
InChI Key
KCDXJAYRVLXPFO-UHFFFAOYSA-N
SMILES
COC1=CC(C=O)=CC(OC)=C1O
Appearance (Color)
White to pale cream to pale brown or pale pink
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Melting Point (clear melt)
109.0-115.0?C

Description

Syringaldehyde has bioactive properties and is, therefore, used in pharmaceuticals, food, cosmetics, textiles, pulp and paper industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Syringaldehyde has bioactive properties and is, therefore, used in pharmaceuticals, food, cosmetics, textiles, pulp and paper industries.

Solubility
Very sparingly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store at room temperature. It is sensitive to air. Incompatible with strong oxidizing agents, and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text