Thermo Scientific Chemicals

4-Hydroxyphenoxyacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12940.22
100 g, Each
Thermo Scientific Chemicals

4-Hydroxyphenoxyacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12940.22
100 g, Each
Quantity
Catalog number: A12940.22
also known as A12940-22
Price (USD)
Price: 863.00
Online price: 746.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1878-84-8
IUPAC Name
2-(4-hydroxyphenoxy)acetate
Molecular Formula
C8H7O4
InChI Key
PKGWLCZTTHWKIZ-UHFFFAOYSA-M
SMILES
OC1=CC=C(OCC([O-])=O)C=C1
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (Silylated GC)
≥98.0%
Melting Point (clear melt)
150.0-158.0°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text