2-Bromo-1,4-difluorobenzene, 98%
2-Bromo-1,4-difluorobenzene, 98%
2-Bromo-1,4-difluorobenzene, 98%
Thermo Scientific Chemicals

2-Bromo-1,4-difluorobenzene, 98%

CAS: 399-94-0 | C6H3BrF2 | 192.991 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12963.22
also known as A12963-22
Price (EUR)
427,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
427,00
Each
Chemical Identifiers
CAS399-94-0
IUPAC Name2-bromo-1,4-difluorobenzene
Molecular FormulaC6H3BrF2
InChI KeyXCRCSPKQEDMVBO-UHFFFAOYSA-N
SMILESFC1=CC=C(F)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5060-1.5100 @ 20?C
FormLiquid
Assay (GC)≥97.5 to ≤102.5%
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only