4-Bromomandelic acid, 98+%
4-Bromomandelic acid, 98+%
4-Bromomandelic acid, 98+%
4-Bromomandelic acid, 98+%
Thermo Scientific Chemicals

4-Bromomandelic acid, 98+%

CAS: 6940-50-7 | C8H7BrO3 | 231.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A12978.14
also known as A12978-14
Price (TWD)
-
Request A Quote
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS6940-50-7
IUPAC Name2-(4-bromophenyl)-2-hydroxyacetic acid
Molecular FormulaC8H7BrO3
InChI KeyBHZBRPQOYFDTAB-UHFFFAOYNA-N
SMILESOC(C(O)=O)C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)114.0-120.0?C
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (GC)≥98.0%
4-Bromomandelic acid is used as a pharmaceutical intermediate. 4-bromomandelic acid is mainly used as an analytical reagent for zirconium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromomandelic acid is used as a pharmaceutical intermediate. 4-bromomandelic acid is mainly used as an analytical reagent for zirconium.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
RUO – Research Use Only

General References:

  1. UC Dyer.; PD Shapland.; PD Tiffin. Preparation of enantiopure 4-arylmandelic acids via a Pd/C catalysed Suzuki coupling of enantiopure 4-bromomandelic acid. Tetrahedron Letters. 200142, (9) , 1765-1767.
  2. MG Schmid.;O Lecnik.; U Sitte.; G Gübitz. Application of ligand-exchange capillary electrophorersis to the chiral separation of α-hydroxyacids and β-blockers. Journal of Chromatography A. 2000875, (1-2) , 307-314.