3-Phenoxybenzyl alcohol, 98%
3-Phenoxybenzyl alcohol, 98%
3-Phenoxybenzyl alcohol, 98%
Thermo Scientific Chemicals

3-Phenoxybenzyl alcohol, 98%

CAS: 13826-35-2 | C13H12O2 | 200.237 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A13008.09
also known as A13008-09
Price (TWD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS13826-35-2
IUPAC Name(3-phenoxyphenyl)methanol
Molecular FormulaC13H12O2
InChI KeyKGANAERDZBAECK-UHFFFAOYSA-N
SMILESOCC1=CC=CC(OC2=CC=CC=C2)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.5905-1.5945 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only