Acridine Orange
Invitrogen™

Acridine Orange

アクリジンオレンジは、dsDNAに結合すると緑色の蛍光を発し、ssDNAまたはRNAに結合すると赤色の蛍光を発する細胞透過性核酸結合色素です。この独自の特性のため、アクリジンオレンジは細胞周期研究に有用です。アクリジンオレンジは、リソソソーム色素としても使用されています。この色素は溶液(A-3568)で提供されます詳細を見る
製品番号(カタログ番号)数量
A13011 g
製品番号(カタログ番号) A1301
価格(JPY)
21,600
キャンペーン価格
Ends: 25-Sep-2026
36,000
割引額 14,400 (40%)
Each
数量:
1 g
アクリジンオレンジは、dsDNAに結合すると緑色の蛍光を発し、ssDNAまたはRNAに結合すると赤色の蛍光を発する細胞透過性核酸結合色素です。この独自の特性のため、アクリジンオレンジは細胞周期研究に有用です。アクリジンオレンジは、リソソソーム色素としても使用されています。この色素は溶液(A-3568)で提供されます。
研究用途にのみご使用ください。診断目的には使用できません。
仕様
オレンジ
検出法蛍光
染色剤タイプ細胞透過性
励起波長域500⁄526(DNA)、460⁄650(RNA)
使用対象 (装置)蛍光顕微鏡
数量1 g
出荷条件室温
標識タイプFluorescent Dye
製品タイプ色素
Sub Cellular Localization核、核酸, Nucleus
Unit SizeEach
組成および保存条件
室温で保存し、光から保護します。

引用および参考文献 (350)

引用および参考文献
Abstract
Low-dose estrogen therapy ameliorates experimental autoimmune encephalomyelitis in two different inbred mouse strains.
Authors:Bebo BF Jr,Fyfe-Johnson A,Adlard K,Beam AG,Vandenbark AA,Offner H
Journal:Journal of immunology (Baltimore, Md. : 1950)
PubMed ID:11160259
Spatial variations in growth rate within Klebsiella pneumoniae colonies and biofilm.
Authors:Wentland EJ,Stewart PS,Huang CT,McFeters GA
Journal:Biotechnology progress
PubMed ID:8652119
The use of acridine orange to visualize and quantify spatial variations in growth rate within Klebsiella pneumoniae colonies and biofilm was investigated. Bacterial colonies supported on polycarbonate filter membranes were grown on R2A agar plates. Some colonies were sampled for cell enumeration, while others were cryoembedded, sectioned, and stained with ... More
Rapid assessment of physiological status in Escherichia coli using fluorescent probes.
Authors:Porter J, Edwards C, Pickup RW
Journal:J Appl Bacteriol
PubMed ID:7592133
'Rapid and direct viability assessment of Escherichia coli in filtered, sterile lake water was possible using multiparameter flow cytometry. Fluorescent dyes were used as probes for different cellular functions (membrane potential, membrane integrity and intracellular enzyme activity), which were correlated with the ability of the cells to respond to nutrient ... More
Defective acidification of intracellular organelles in cystic fibrosis.
Authors:Barasch J, Kiss B, Prince A, Saiman L, Gruenert D, al-Awqati Q
Journal:Nature
PubMed ID:1712081
'The phenotype of cystic fibrosis (CF) includes abnormalities in transepithelial transport of Cl- (refs 1-5), decreased sialylation and increased sulphation and fucosylation of glycoproteins, and lung colonization with Pseudomonas. It is not apparent how these abnormalities are interrelated, nor how they result from loss of function of the CF gene-encoded ... More
Rapid movements of vimentin on microtubule tracks: kinesin-dependent assembly of intermediate filament networks.
Authors:Prahlad V, Yoon M, Moir RD, Vale RD, Goldman RD
Journal:J Cell Biol
PubMed ID:9763428
'The assembly and maintenance of an extended intermediate filament (IF) network in fibroblasts requires microtubule (MT) integrity. Using a green fluorescent protein-vimentin construct, and spreading BHK-21 cells as a model system to study IF-MT interactions, we have discovered a novel mechanism involved in the assembly of the vimentin IF cytoskeleton. ... More