This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
3-Phenyl-1-propanol has a natural fragrance with antimicrobial properties against bacteria and molds. It is used in combination with Heliotropin or Piperonal as a preservative for cosmetic products. Its ester derivatives are used as fragrance component in fresh flower compositions, like lilac, hyacinth and lily of the valley because of the balsamic odor character. It is also a useful biochemical for proteomics research.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
General References:
- Soyeong Kang; Hyeon-Kyu Lee. Highly Efficient, Enantioselective Syntheses of (S)-(+)- and (R)-(-)-Dapoxetine Starting with 3-Phenyl-1-propanol. J. Org. Chem. 2010, 75, (1),237-240
- Ignacio Medina; Julio L.Bueno. Solubilities of 2-Nitroanisole and 3-Phenyl-1-propanol in Supercritical Carbon Dioxide. J. Chem. Eng. Data. 2000, 45, (2),298-300