1-Pentanol is used as a key starting material in the production of dichloro-acetic acid pentyl ester, 2-methyltetrahydrofuran, di-n-penyl ether (DNPE), pentyl butyrate and amyl acetate. As a solvent, it is used for coating CDs and DVDs. It has been considered to be a replacement for gasoline and diesel fuels due to its combustion properties. Further, it is used as a reagent to prepare derivatives of ferulic acid and caffeic acid, which shows anticancer properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Pentanol is used as a key starting material in the production of dichloro-acetic acid pentyl ester, 2-methyltetrahydrofuran, di-n-penyl ether (DNPE), pentyl butyrate and amyl acetate. As a solvent, it is used for coating CDs and DVDs. It has been considered to be a replacement for gasoline and diesel fuels due to its combustion properties. Further, it is used as a reagent to prepare derivatives of ferulic acid and caffeic acid, which shows anticancer properties.
Solubility
Miscible with water, alcohol, ether, acetone and most organic solvents.
Notes
Incompatible with strong oxidizing agents, alkali metals, strong acids, halides and aldehydes.
RUO – Research Use Only
General References:
- Estrada-Baltazar, A.; Bravo-Sanchez, M. G.; Iglesias-Silva, G. A.; Alvarado, J. F. J.; Castrejon-Gonzalez, E. O.; Ramos-Estrada, M. Densities and viscosities of binary mixtures of n-decane+ 1-pentanol,+ 1-hexanol,+ 1-heptanol at temperatures from 293.15 to 363.15 K and atmospheric pressure. Chin. J. Chem. Eng. 2015, 23 (3), 559-571.
- Khoiroh, I.; Shu, S. S.; Lee, M. J. Vapor-liquid equilibrium of polyethylene glycol monooleyl ether with 2-butanol, tert-butanol, or 1-pentanol. Fluid Phase Equilib. 2015, 404, 81-88.