2-Bromo-m-xylene has been used to prepare 2,2,4,6,6-pentamethylbiphenyl. It is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El 2-bromo-m-xileno se ha utilizado para preparar 2,2',4,6,6'-pentametilbifenilo. Se utiliza como intermediario en la síntesis orgánica.
Solubilidad
Soluble en solución acuosa. Muy soluble en etanol, soluble en acetona y benceno.
Notas
Estable en las condiciones de almacenamiento recomendadas. Incompatible con agentes oxidantes. Debe almacenarse en un recipiente herméticamente cerrado. Almacenar en una zona fresca, seca y bien ventilada.
RUO – Research Use Only
- Andreas Schmidt; Alireza Rahimi. A versatile catalyst system for Suzuki-Miyaura syntheses of sterically hindered biaryls employing a cyclobutene-1,2-bis(imidazolium) salt. Chemical Communications. 2010, 46, (17),2995-2997