2-Bromo-m-xylene has been used to prepare 2,2,4,6,6-pentamethylbiphenyl. It is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-m-xylene has been used to prepare 2,2′,4,6,6′-pentamethylbiphenyl. It is used as an intermediate in organic synthesis.
Solubility
Soluble in aqueous solution. Very soluble in ethanol, soluble in acetone and benzene.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents. Should be stored in a tightly closed container. Store in a cool, dry, well-ventilated area.
RUO – Research Use Only
Andreas Schmidt; Alireza Rahimi. A versatile catalyst system for Suzuki-Miyaura syntheses of sterically hindered biaryls employing a cyclobutene-1,2-bis(imidazolium) salt. Chemical Communications. 2010, 46, (17),2995-2997