Thermo Scientific Chemicals

N-(9-Fluorenylmethoxycarbonyloxy)succinimide, 98%, Thermo Scientific Chemicals

Catalog number: A13143.03
1 g, Each
Thermo Scientific Chemicals

N-(9-Fluorenylmethoxycarbonyloxy)succinimide, 98%, Thermo Scientific Chemicals

Catalog number: A13143.03
1 g, Each
Quantity
Catalog number: A13143.03
also known as A13143-03
Price (USD)
Price: 31.70
Online price: 27.65
Your price:
Quantity
-

Chemical Identifiers

CAS
82911-69-1
IUPAC Name
2,5-dioxopyrrolidin-1-yl (9H-fluoren-9-yl)methyl carbonate
Molecular Formula
C19H15NO5
InChI Key
WMSUFWLPZLCIHP-UHFFFAOYSA-N
SMILES
O=C(OCC1C2=CC=CC=C2C2=CC=CC=C12)ON1C(=O)CCC1=O
Appearance (Color)
White
Form
Powder
Assay (HPLC)
≥97.5%

Description

N-(9-Fluorenylmethoxycarbonyloxy)succinimide is the most efficient reagent for the selective preparation of N-(9-fluorenylmethoxycarbonyl) derivatives of hydroxy-amino acids. Dipeptide formation is lower than with the chloroformate, Fmoc-Cl. It is also employed in the synthesis of glycopeptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(9-Fluorenylmethoxycarbonyloxy)succinimide is the most efficient reagent for the selective preparation of N-(9-fluorenylmethoxycarbonyl) derivatives of hydroxy-amino acids. Dipeptide formation is lower than with the chloroformate, Fmoc-Cl. It is also employed in the synthesis of glycopeptides.

Solubility
Soluble in dimethyl sulfoxide and dimethyl formamide.

Notes
Store in a cool place. Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text