Use with Annexin Binding Buffer for flow cytometry
Annexin V Conjugates for Apoptosis Detection
Invitrogen™

Annexin V Conjugates for Apoptosis Detection

Detect early stages of apoptosis with Annexin V stand-alone Alexa Fluor, APC, Pacific Blue, PE, FITC, and biotin conjugates using flow cytometry.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberExcitation/EmissionFlow Cytometer Laser LinesConjugate
A13204Biotin-X
A13201495/519488Alexa Fluor 488
A13199494/518488FITC
A13202578/603532, 561Alexa Fluor 568
A13203590/617532Alexa Fluor 594
A23202346/442UVAlexa Fluor 350
A23204650/665633-637Alexa Fluor 647
A35108555/565532, 561Alexa Fluor 555
A35109679/702633-637Alexa Fluor 680
A35110650/660633-637APC (Allophycocyanin)
A35111565/578488, 532, 561PE
A35122410/455405Pacific Blue
Catalog number A13204
Price (USD)
498.00
Each
Conjugate:
Biotin-X
Price (USD)
498.00
Each
Achieve quick and reliable detection of early cell apoptosis with Annexin V stand-alone conjugates for apoptosis detection. Annexin V conjugates offer up to 100-fold difference in fluorescence signal intensity between apoptotic and non-apoptotic cells using flow cytometry.
Annexin V has a high affinity for phosphatidylserine (PS), which becomes exposed on the outer leaflet of cells undergoing apoptosis. Because of this affinity, fluorescently labeled annexin V reagents are commonly used in apoptosis research.

Annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, an indicator of intermediate stages of apoptosis. The difference in fluorescence intensity between apoptotic and nonapoptotic cells stained with our fluorescent annexin V conjugates, as measured by flow cytometry, is typically about 100-fold.

In collaboration with Nexins Research BV, we provide the best and brightest annexin V conjugates available, including Alexa Fluor 350, 488, 555, 568, 594, 647, and 680 annexin V conjugates, as well as Annexin V APC, Biotin-X, FITC, Pacific Blue, and PE conjugates. Highly fluorescent annexin V conjugates provide quick and reliable detection methods for studying the externalization of phosphatidylserine, one of the earliest indicators of apoptosis.

The Annexin V Pacific Blue conjugate is violet excitable, making it ideal for instruments with a violet laser and for multicolor experiments that include green- or red-fluorescent dyes.

The benefits of our annexin V conjugates include:
• Conjugated to Invitrogen Alexa Fluor and eFluor dyes for brighter signals
• Conjugates for all available lasers
• Available as stand-alone reagents or easy-to-use kits

Annexin V staining to detect apoptotic cells can only be done on live cells and tissue. If samples are to be fixed post-staining, there are specific conditions required to achieve transient retention of signal. These include use of an alcohol-free, aldehyde-based fixation method, use of buffers containing Ca2+ and avoidance of surfactants/detergents. For your convenience, we also offer a concentrated annexin-binding buffer that facilitates the binding of annexin V to phosphatidylserine in apoptosis assays.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionAnnexin V, biotin-X conjugate
For Use With (Equipment)Flow Cytometer
No. of Reactions100
Product TypeAnnexin V conjugate
Quantity500 μL
Shipping ConditionWet Ice
ConjugateBiotin-X
Unit SizeEach
Contents & Storage
Contains 1 vial of annexin V, biotin-x conjugate. Store in refrigerator (2°C to 8°C) and protect from light.

Frequently asked questions (FAQs)

I want to study apoptosis using an Annexin V conjugate, but with adherent cells via microscopy instead of flow cytometry. Can this be done?

It has been done, but we don‘t recommend it. Both healthy cells and apoptotic cells possess phosphatidylserine on the cell surface, which can be detected with Annexin V, but apoptotic cells have significantly more of it. You can easily tell the difference between these two populations with flow cytometry, because flow cytometers are more sensitive and have a higher throughput. But with a microscope, you cannot always tell the difference, especially for adherent cells. Instead, for microscopy, we recommend a different technique, such as detecting caspases with CellEvent Caspase Detection Reagents.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

When should I use Annexin V instead of an Invitrogen CellEvent Caspase Detection Reagent?

Choose Annexin V to detect phosphatidylserine exposure at the plasma membrane, and choose Invitrogen CellEvent Caspase Detection Reagents to detect activated caspases and nuclear fluorescence. The methods measure different apoptotic events and should not be treated as interchangeable.

I am trying to label adherent cells with annexin V and am finding that everything is getting labeled. How can I fix this?

Treating cells with trypsin or other reagents to detach adherent cells causes damage to the membrane, such that cells will be labeled with annexin V. The best way to avoid this problem is to allow your cells to recover for 30-45 min in the incubator. Swirl the tube/plate/flask every few minutes to prevent re-attachment. After this recovery period, you can label your cells with annexin V and analyze by flow cytometry.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What are the advantages of flow cytometry?

-Measures data from single cells.
-Data are obtained for a large number of cells, generating a rich statistical analysis of cell populations.
-Because single cells are measured, it will reveal heterogeneity within a population.
-With the ability to multiplex, small sub-populations can be identified.
-Thousands of cells can be analyzed rapidly.
-It is ideally suited for blood samples and other cells in suspension.
-Data can be re-analyzed multiple times after acquisition.
-Flow cytometry files (FCS) can be archived.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What kinds of applications can I run on a flow cytometer?

There are several applications, some of which include immunophenotyping, cell cycle analysis, apoptosis assays such as annexin V staining, CellEvent Caspase-3/7 assay, and TUNEL assay, cell viability, proliferation assays such as CellTrace assay and Click-iT EdU assay, measurements of mitochondrial potential with MitoProbe assays, and cell counting using counting beads.

Find additional tips, troubleshooting help, and resources within our Flow Cytometry Support Center.

Citations & References (14)

Citations & References
Abstract
Regulation of the Fas death pathway by FLICE-inhibitory protein in primary human B cells.
Authors:Hennino A, Berard M, Casamayor-Pallejà M, Krammer PH, Defrance T
Journal:J Immunol
PubMed ID:10975811
'The Fas/Fas ligand (L) system plays an important role in the maintenance of peripheral B cell tolerance and the prevention of misguided T cell help. CD40-derived signals are required to induce Fas expression on virgin B cells and to promote their susceptibility to Fas-mediated apoptosis. In the current study, we ... More
Detection of apoptotic cells by annexin V labeling at electron microscopy.
Authors:Pellicciari C, Bottone MG, Biggiogera M
Journal:Eur J Histochem
PubMed ID:9359032
'Annexin V is a Ca(++)-binding protein which is widely used as a marker for apoptotic cells, as it binds to the phosphatidylserine residues exposed at the surface of apoptotic cells. In this paper, we describe a method for the immunogold-labeling of biotin-conjugated Annexin V, to detect apoptotic thymocytes at electron ... More
Spatiotemporal distribution of dying neurons during early mouse development.
Authors:van den Eijnde SM, Lips J, Boshart L, Vermeij-Keers C, Marani E, Reutelingsperger CP, De Zeeuw CI
Journal:Eur J Neurosci
PubMed ID:10051772
'Apoptosis is a critical cellular event during several stages of neuronal development. Recently, we have shown that biotinylated annexin V detects apoptosis in vivo in various cell lineages of a wide range of species by binding to phosphatidylserines that are exposed at the outer leaflet of the plasma membrane. In ... More
In situ detection of apoptosis during embryogenesis with annexin V: from whole mount to ultrastructure.
Authors:van den Eijnde SM, Luijsterburg AJ, Boshart L, De Zeeuw CI, van Dierendonck JH, Reutelingsperger CP, Vermeij-Keers C
Journal:Cytometry
PubMed ID:9415414
'Apoptosis is of paramount importance during embryonic development. This insight stems from early studies which correlated cell death to normal developmental processes and now has been confirmed by linking aberrant cell death patterns to aberrant development. Linking apoptosis to the phenotype of a developing organism requires spatial information on the ... More
Complement-independent, peroxide-induced antibody lysis of platelets in HIV-1-related immune thrombocytopenia.
Authors:Nardi M, Tomlinson S, Greco MA, Karpatkin S
Journal:Cell
PubMed ID:11551503
'Immunologic thrombocytopenia is seen commonly in HIV-1 infection. The pathogenesis of this problem has been unclear, but it is associated with circulating immune complexes that contain platelet membrane components and anti-platelet membrane GPIIIa49-66 IgG antibodies. These antibodies cause acute thrombocytopenia when injected into mice. We now show that purified anti-GPIIIa49-66 ... More