Thermo Scientific Chemicals

2-Bromopyridine, 99%, Thermo Scientific Chemicals

Catalog number: A13241.14
25 g, Each
Thermo Scientific Chemicals

2-Bromopyridine, 99%, Thermo Scientific Chemicals

Catalog number: A13241.14
25 g, Each
Quantity
Catalog number: A13241.14
also known as A13241-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
109-04-6
IUPAC Name
2-bromopyridine
Molecular Formula
C5H4BrN
InChI Key
IMRWILPUOVGIMU-UHFFFAOYSA-N
SMILES
BrC1=CC=CC=N1
Appearance (Color)
Clear colorless to pale yellow or pale brown
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Refractive Index
1.5695-1.5725 @ 20?C

Description

2-Bromopyridine is used in the preparation of a variety of biologically active compounds like antimalarial agents. It is also used to prepare beta-adrenoceptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopyridine is used in the preparation of a variety of biologically active compounds like antimalarial agents. It is also used to prepare beta-adrenoceptor agonist.

Solubility
Slightly miscible with water.

Notes
Keep away from heat, sparks, and flame. Hygroscopic. Incompatible with strong oxidizing agents, strong acids and acid chlorides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text