Bis(tri-n-butyltin) oxide, 97%
Bis(tri-n-butyltin) oxide, 97%
Bis(tri-n-butyltin) oxide, 97%
Thermo Scientific Chemicals

Bis(tri-n-butyltin) oxide, 97%

CAS: 56-35-9 | C24H58OSn2 | 600.15 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A13242.0E
または、製品番号A13242-0E
価格(JPY)
-
数量:
2500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS56-35-9
IUPAC Nametributyltin oxide
Molecular FormulaC24H58OSn2
InChI KeyNZQFYJHTWVEUPP-UHFFFAOYSA-N
SMILESO.CCCC[SnH](CCCC)CCCC.CCCC[SnH](CCCC)CCCC
さらに表示
Appearance (Color)Colorless to pale yellow
FormLiquid
Assay from Supplier's CofA≥96%
Bis(tri-n-butyltin) oxide is employed in the synthesis of α,β-unsaturated methyl ketones, isoxazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(tri-n-butyltin) oxide is employed in the synthesis of α,β-unsaturated methyl ketones, isoxazoles.

Solubility
Soluble in water (0.071 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
RUO – Research Use Only

General References:

  1. E.I. Krajnc.; P.W. Wester.;J.G. Loeber.; F.X.R. van Leeuwen.; J.G. Vos.; H.A.M.G. Vaessen.; C.A. van der Heijden. Toxicity of bis(tri-n-butyltin)oxide in the rat: I. Short-term effects on general parameters and on the endocrine and lymphoid systems. Toxicol. Appl. Pharmacol. 1984, 75 (3),363-386 .
  2. J.G. Vos.; A.De Klerk.; E.I. Krajnc.; H.Van Loveren.; J. Rozing. Immunotoxicity of Bis(tri-n-butyltin)oxide in the rat: Effects on thymus-dependent immunity and on nonspecific resistance following long-term exposure in young versus aged rats. Toxicol. Appl. Pharmacol. 1990, 105 (1),144-155 .
  3. Reacts with alcohols to give tri-n-butylstannyl ethers which, for simple alcohols, are very susceptible to hydrolysis: Synthesis, 56 (1969); J. Organomet. Chem., 110, C57 (1976). In combination with bromine or NBS, allylic, benzylic and secondary alcohols are oxidized to carbonyl compounds, enabling the selective oxidation of secondary, in the presence of primary, alcohols: Bull. Chem. Soc. Jpn., 49, 1656 (1976); Tetrahedron Lett., 4597 (1976); J. Am. Chem. Soc., 98, 1629 (1976). Similarly, sulfides give sulfoxides with no over-oxidation to sulfones: Tetrahedron Lett., 2413 (1977).
  4. Reacts with terminal alkynes to give alkynyl tin reagents. Where these reagents have electron-withdrawing substituents, they undergo regioselective cycloaddition with 1,3-dienes: Tetrahedron, 45, 1145 (1989):
  5. Converts thioamides to nitriles, as does Di-n-butyl tin oxide, L14491: J .Org. Chem., 47, 4594 (1982).
  6. Reagent for mild, selective non-hydrolytic deprotection of esters: Tetrahedron Lett., 32, 4239 (1991); J. Org. Chem., 59, 7259 (1994); Tetrahedron Lett., 36, 3311 (1995).