PrestoBlue™ and PrestoBlue™ HS Cell Viability Reagents
PrestoBlue™ and PrestoBlue™ HS Cell Viability Reagents
PrestoBlue™ and PrestoBlue™ HS Cell Viability Reagents
Invitrogen™

PrestoBlue™ and PrestoBlue™ HS Cell Viability Reagents

Monitor cell proliferation and viability with the resazurin-based PrestoBlue and PrestoBlue HS cell viability reagents. These nontoxic and cell permeable reagents quantitatively measure cell viability in as little as 10 minutes using absorbance- or fluorescence-based microplate readers.

Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidadGrado de pureza o calidad
A1326125 mLEstándar
A13262100 mLEstándar
P5020025 mlHS
P50201100 mlHS
Número de catálogo A13261
Precio (MXN)
8,237.87
Each
Cantidad:
25 mL
Grado de pureza o calidad:
Estándar
Precio (MXN)
8,237.87
Each
PrestoBlue and PrestoBlue HS cell viability reagents are ready-to-use, nontoxic, resazurin-based solutions that function as long term kinetic cell health indicators for fluorescence and absorbance-based microplate assays. These reagents are cell permeable and use the reducing power of living cells to quantitatively measure cell proliferation and viability in as little as 10 minutes. PrestoBlue HS Cell Viability Reagent is an improved version of PrestoBlue reagent, containing highly purified resazurin, which increases sensitivity and reduces background in cell proliferation assays. Add the power of multiplexing with reagents such as the CyQUANT Direct assay to improve your data analysis.

Measuring changes in cell viability is a fundamental method for assessing cell health, determining genotoxicity, and evaluating anti-cancer drugs. Cell health can be monitored by detecting changes in several key indicators, including changes to plasma membrane integrity, DNA synthesis, DNA content, enzyme activity, presence of ATP, and cellular reducing environment.

Features of PrestoBlue and PrestoBlue HS cell viability reagents include:

  • Optimized for reliable detection of mammalian cell viability in only 10 minutes
  • Measure cell viability and proliferation in various cell types, including mammalian (2D and 3D), bacteria, and yeast cells
  • A convenient add-and-read format—no mixing, washing, or cell lysis required
  • Compatible with either fluorescence- or absorbance-based microplate readers
  • Obtain high quality cell proliferation results with a large dynamic range
  • Monitor living cells using real time kinetics and continue culturing them for downstream multiplexed functional assays

Quick 10-minute incubations

Detect mammalian cell viability after just a 10-minute incubation with PrestoBlue and PrestoBlue HS cell viability reagents. These are the only live-cell viability reagents on the market that provide robust data in 10 minutes following reagent addition. All other resazurin-based cell viability reagents require a 1–4 hour incubation with the reagent prior to obtaining data. For best results using 3D samples, it is recommended to incubate the samples for 18–24 hours prior to measurements.

Exceptional sensitivity and accuracy

PrestoBlue and PrestoBlue HS cell viability reagents can detect as few as 12 cells per well in a 384-well plate, depending on the cell type and incubation time. After a 10-minute incubation, PrestoBlue reagent can linearly detect cells down to 98 cells/well with a Z' value greater than 0.5.

Simplified assay format

PrestoBlue and PrestoBlue HS cell viability reagents are addition-only reagents, eliminating the need for cell lysis or solubilization steps required by other assays. This minimizes hands-on time and simplifies the process.

Nontoxic live cell assays

PrestoBlue and PrestoBlue HS cell viability reagents are nontoxic and do not require cell lysis, allowing cells to be incubated for up to 24 hours. Since no lysis is required, the diluted PrestoBlue or PrestoBlue HS solution can be removed from the cells and replaced with complete growth medium for further culturing and use in downstream assays.

PrestoBlue HS reagent—a highly purified version of PrestoBlue reagent with improved performance

All resazurin-based reagents contain a detectable amount of resorufin contamination due to the synthesis and manufacturing process, which contributes to high background fluorescence and reduces performance in cell viability assays. To reduce background and improve performance, an innovative purification process was developed to remove the contaminating resorufin, resulting in the highly pure resazurin used in the PrestoBlue HS Cell Viability Reagent. This highly purified, nontoxic resazurin reduces background fluorescence by >50% and increases signal-to-background by >100% in cell viability microplate assays.

Features of PrestoBlue HS Cell Viability Reagent include:

  • Removal of contaminants from resazurin, resulting in >50% reduction in background fluorescence
  • Increased signal-to-background ratio by >100%, providing a large assay signal window
  • Offers high sensitivity with a linear response, detecting as few as 10 cells per well

Mechanism of PrestoBlue and PrestoBlue HS reagents

Using resazurin-based reagents to monitor changes in cellular reducing environment or metabolic activity is a well-established and reliable method to indicate cell viability or death. PrestoBlue and PrestoBlue HS reagents contain resazurin, which is a nontoxic, cell permeable compound that is blue in color and virtually nonfluorescent. When added to cells, the resazurin-based PrestoBlue and PrestoBlue HS reagents enter cells and are modified by the reducing environment of viable cells. In live cells, resazurin is reduced to resorufin, a compound that is red in color and highly fluorescent. Viable cells continuously convert resazurin to resorufin, increasing the overall fluorescence of the medium surrounding the cells. The conversion of resazurin to resorufin also results in a pronounced color change, allowing cell viability to be detected using either absorbance- or fluorescence-based plate readers.

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
Tipo de célulaBacterias, células eucariotas, células fúngicas
Concentración10 células por pocillo (placa de 384 pocillos)
Método de detecciónAbsorbance, Fluorescence
Tipo de coloranteResazurina
Longitud de onda de emisión590-615 nm
Longitud de onda de excitación535-560 nm
FormularioLíquido
FormatoCuvettes, 96-well Plate, 384-well Plate
Tiempo de incubaciónDe 10 min a 2 h
Grado de pureza o calidadEstándar
Cantidad25 mL
Condiciones de envíoAprobado para su envío a temperatura ambiente o en hielo húmedo
Capacidad de procesamientoCompatible con alto rendimiento
Absorbance Wavelength570 nm
ColorAzul
Intervalo de longitud de onda de emisión585 to 600 nm
Excitation Wavelength Range555 to 570 nm
Para utilizar con (aplicación)Cell Viability
Para utilizar con (equipo)Lector de microplacas, Espectrofotómetro, Lector HTS
Línea de productosPrestoBlue
Tipo de productoReactivo
Reference Wavelength600 nm
Unit SizeEach
Contenido y almacenamiento
Almacenar a 4 °C y proteger de la luz.

Preguntas frecuentes

MTT cell proliferation plate assays require cellular metabolism to modify the reagent and thus, only live cells will be counted. Is this true for CyQUANT Direct Cell Proliferation Assay, too?

This is true for MTT (as well as XTT, AlamarBlue Cell Viability Reagent, and PrestoBlue Cell Viability Reagent). CyQUANT Direct will also only count live cells, but for a different reason. The dye in it is a green-fluorescent nucleic acid stain, which will bind to DNA in all cells, live or dead, without the need for cellular metabolism. However, there is a non-cell-permeable quenching reagent in the kit which will both quench extracellular fluorescence (and thus this is a no-wash assay) AND will quench the fluorescence in dead cells (but not live cells).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

MTT cell proliferation assays require cellular metabolism to turn over the reagent, and thus only live cells will be counted. Is this true for CyQUANT Direct as well?

This is true for MTT (as well as XTT, AlamarBlue Cell Viability Reagent, and PrestoBlue Cell Viability Reagent). CyQUANT Direct will also only count live cells, but for a different reason. The dye is a green-fluorescent nucleic acid stain, which will bind to DNA in all cells, live or dead, without the need for cellular metabolism. However, there is a cell impermeant quenching reagent in the kit which will quench both extracellular fluorescence (and thus this is a no-wash assay) and the fluorescence in dead cells (but not live cells).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I combine Invitrogen CellEvent Caspase-3/7 Green Detection Reagent with PrestoBlue Cell Viability Reagent?

Yes. Invitrogen CellEvent Caspase-3/7 Green Detection Reagent has been demonstrated in combination with Invitrogen PrestoBlue Cell Viability Reagent in the same sample. CellEvent Caspase reagent fluorescence reports activated caspase, while PrestoBlue Cell Viability Reagent reports cellular metabolic activity as an indicator of viability. Follow the instructions for each reagent and use single-reagent controls to evaluate signal interference.

Why would I use PrestoBlue HS Cell Viability Reagent instead of the original PrestoBlue Cell Viability Reagent?

PrestoBlue HS Cell Viability Reagent provides a lower background, suitable for analyzing a lower number of cells per well. Also, the removal of impurities in the 'HS' reagent limits any artifacts that may be critical to your cells' viability.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Does PrestoBlue HS Cell Viability Reagent have the same properties as the original PrestoBlue Cell Viability Reagent?

Yes and in addition, PrestoBlue HS Cell Viability Reagent has a higher purity which results in a lower background.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (51)

Citations & References
Abstract
Portable self-contained cultures for phage and bacteria made of paper and tape.
Authors:Funes-Huacca M, Wu A, Szepesvari E, Rajendran P, Kwan-Wong N, Razgulin A, Shen Y, Kagira J, Campbell R, Derda R,
Journal:Lab Chip
PubMed ID:22895550
In this paper, we demonstrate that a functional, portable device for the growth of bacteria or amplification of bacteriophage can be created using simple materials. These devices are comprised of packing tape, sheets of paper patterned by hydrophobic printer ink, and a polydimethyl siloxane (PDMS) membrane, which is selectively permeable ... More
Galangin inhibits tumour growth and metastasis of B16F10 melanoma.
Authors:Zhang W, Tang B, Huang Q, Hua Z,
Journal:J Cell Biochem
PubMed ID:22887049
Galangin, an active flavonoid extracted from the root of the Alpinia officinarum Hance, showed a cytotoxic effect on several cancer cell lines in vitro. However, there is no information available concerning its antimetastatic effect. Focal adhesion kinase (FAK), a cytoplasmic tyrosine kinase, involves in many aspects of cellular processes such ... More
Efficient replication of genotype 3a and 4a hepatitis C virus replicons in human hepatoma cells.
Authors:Saeed M, Scheel TK, Gottwein JM, Marukian S, Dustin LB, Bukh J, Rice CM,
Journal:Antimicrob Agents Chemother
PubMed ID:22869572
Despite recent advances in the treatment of hepatitis C, the quest for pan-genotype, effective, and well-tolerated inhibitors continues. To facilitate these efforts, it is desirable to have in vitro replication systems for all major HCV genotypes. However, cell culture replication systems exist for only genotypes 1a, 1b, and 2a. In ... More
Suppressor of Cytokine Signaling 6 (SOCS6) Negatively Regulates Flt3 Signal Transduction through Direct Binding to Phosphorylated Tyrosines 591 and 919 of Flt3.
Authors:Kazi JU, Sun J, Phung B, Zadjali F, Flores-Morales A, Rönnstrand L,
Journal:J Biol Chem
PubMed ID:22952242
The receptor tyrosine kinase Flt3 is an important growth factor receptor in hematopoiesis, and gain-of-function mutations of the receptor contribute to the transformation of acute myeloid leukemia. SOCS6 (suppressor of cytokine signaling 6) is a member of the SOCS family of E3 ubiquitin ligases that can regulate receptor tyrosine kinase ... More
a-Synuclein Induces Alterations in Adult Neurogenesis in Parkinson Disease Models via p53-mediated Repression of Notch1.
Authors:Desplats P, Spencer B, Crews L, Pathel P, Morvinski-Friedmann D, Kosberg K, Roberts S, Patrick C, Winner B, Winkler J, Masliah E,
Journal:J Biol Chem
PubMed ID:22833673
Parkinson disease is characterized by the loss of dopaminergic neurons mainly in the substantia nigra. Accumulation of a-synuclein and cell loss has been also reported in many other brain regions including the hippocampus, where it might impair adult neurogenesis, contributing to nonmotor symptoms. However, the molecular mechanisms of these alterations ... More