3-(Chloromethyl)pyridine hydrochloride, 97%
3-(Chloromethyl)pyridine hydrochloride, 97%
3-(Chloromethyl)pyridine hydrochloride, 97%
3-(Chloromethyl)pyridine hydrochloride, 97%
Thermo Scientific Chemicals

3-(Chloromethyl)pyridine hydrochloride, 97%

CAS: 6959-48-4 | C6H7Cl2N | 164.029 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A13279.22
also known as A13279-22
Price (TWD)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS6959-48-4
IUPAC Namehydrogen 3-(chloromethyl)pyridine chloride
Molecular FormulaC6H7Cl2N
InChI KeyUZGLOGCJCWBBIV-UHFFFAOYSA-N
SMILES[H+].[Cl-].ClCC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormCrystalline powder or powder or needles
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Assay (Freebase GC)≥96.0%
Water Content (Karl Fischer Titration)<1.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only