2-Methylbenzyl chloride, 98+%
2-Methylbenzyl chloride, 98+%
2-Methylbenzyl chloride, 98+%
2-Methylbenzyl chloride, 98+%
Thermo Scientific Chemicals

2-Methylbenzyl chloride, 98+%

CAS: 552-45-4 | C8H9Cl | 140.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number A13448.22
also known as A13448-22
Price (EUR)
56,00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (EUR)
56,00
Each
Add to cart
Chemical Identifiers
CAS552-45-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.0%
FormLiquid
Refractive Index1.5395-1.5435 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only