Dexamethasone, 10 mM in DMSO
Gibco™

Dexamethasone, 10 mM in DMSO

This solution contains 10 mM dexamethasone in DMSO. Dexamethasone is an anti-inflammatory glucocorticoid used as a supplement in hepatocyte cellRead more
Have Questions?
Catalog NumberQuantity
A1344950 μL
Catalog number A13449
Price (MXN)
-
Quantity:
50 μL
This solution contains 10 mM dexamethasone in DMSO. Dexamethasone is an anti-inflammatory glucocorticoid used as a supplement in hepatocyte cell culture. This product is available as a separate component for the purposes of supplementing William's E Medium, no phenol red (Cat. No. A1217601) for the maintenance of human spheroid-qualified hepatocytes in 3D.

Note: Refer to the user guide for the human spheroid-qualified hepatocytes for guidance on the formulation of maintenance media for 3D hepatic spheroids.

Specifications
Cell TypeHepatocyte Cells
Concentration10 mM
For Use With (Application)Cell Culture
Quantity50 μL
Shelf Life13 Months from date of manufacture
FormLiquid
Product TypeDMSO
Without AdditivesNo Phenol Red
Unit SizeEach
Contents & Storage
Store in refrigerator (2 to 8°C) and protect from light