Thermo Scientific Chemicals

3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A13594.30
250 g, Each
Thermo Scientific Chemicals

3-Nitrobenzaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: A13594.30
250 g, Each
Quantity
Catalog number: A13594.30
also known as A13594-30
Price (USD)
Price: 60.40
Online price: 51.65
Your price:
Quantity
-

Chemical Identifiers

CAS
99-61-6
IUPAC Name
3-nitrobenzaldehyde
Molecular Formula
C7H5NO3
InChI Key
ZETIVVHRRQLWFW-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=CC(C=O)=C1
Appearance (Color)
Pale cream to cream to pale yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Free acid (titration)
≤0.75%

Description

3-Nitrobenzaldehyde is used in the synthesis of other organic compounds, ranging from pharmaceuticals to plastic additives and intermediate for the processing of perfume and flavoring compounds and in the preparation of certain aniline dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrobenzaldehyde is used in the synthesis of other organic compounds, ranging from pharmaceuticals to plastic additives and intermediate for the processing of perfume and flavoring compounds and in the preparation of certain aniline dyes.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text