o-Toluidine, 99%, Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
o-Toluidine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

o-Toluidine, 99%, Thermo Scientific Chemicals

Have Questions?
產品號碼Quantity
A13602.0B
亦稱為 A13602-0B
1000g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 A13602.0B
亦稱為 A13602-0B
價格 (TWD)
-
Quantity:
1000g
Request bulk or custom format
化學識別
CAS95-53-4
IUPAC Name2-methylaniline
Molecular FormulaC7H9N
InChI KeyRNVCVTLRINQCPJ-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1N
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless or yellow to orange or pink to pale brown
Assay (GC)≥98.5%
FormLiquid
Refractive Index1.5710-1.5740 @ 20?C
o-Toluidine is used in the production of dyes, as a presumptive test for blood in forensic science. And it is also used in determination of glucose in biological materials, as a precursor in the synthesis of poly(o-toluidine) and copper nanoparticle composite material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Toluidine is used in the production of dyes, as a presumptive test for blood in forensic science. And it is also used in determination of glucose in biological materials, as a precursor in the synthesis of poly(o-toluidine) and copper nanoparticle composite material.

Solubility
Soluble in water(15g/L).

Notes
Air & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Aulis Hyvarinen.; Esko A. Nikkila. Specific determination of blood glucose with o-toluidine. Clinica Chimica Acta. 1962, 7, (1), 140-143.

AL-Othmana, Z.A.; Mu. Naushad.; Inamuddin. Organic-inorganic type composite cation exchanger poly-o-toluidine Zr(IV) tungstate: Preparation, physicochemical characterization and its analytical application in separation of heavy metals. Chemical Engineering Journal. 2011, 72, (1), 369-375.