Thermo Scientific Chemicals

4-Methyl-2-nitroaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A13666.0E
2500 g, Each
Thermo Scientific Chemicals

4-Methyl-2-nitroaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A13666.0E
2500 g, Each
Quantity
Catalog number: A13666.0E
also known as A13666-0E
Price (USD)
Price: 260.00
Online price: 221.65
Your price:
Quantity
-

Chemical Identifiers

CAS
89-62-3
Assay (GC)
≥98.0%
Form
Powder
Identification (FTIR)
Conforms
Appearance (Color)
Orange
Melting Point (clear melt)
108.0-118.0°C

Description

4-Methyl-2-nitroaniline is used as a red azo dye.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-2-nitroaniline is used as a red azo dye.

Solubility
Soluble in water (0.2 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text